Assay Details
| Gene Symbol: Sohlh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034961.1 | NP_001030133.1 | 5 - 6 | 563 | 103 | |
| GenBank mRNA | BC104697.1 | - | 5 - 6 | 563 | 7113652 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Sohlh2
Genomic Map