Assay Details
| Gene Symbol: Sohlh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034961.1 | NP_001030133.1 | 6 - 7 | 668 | 135 | |
| GenBank mRNA | BC104697.1 | - | 6 - 7 | 668 | 7113652 | 135 |
More Information
Updated Mapping Information
Set Membership: |
|

Sohlh2
Genomic Map