Assay Details
| Gene Symbol: P2ry12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232408.2 | XP_006232470.1 | 1 - 2 | 1477 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232408.2 transcripts.
Set Membership: |
|

P2ry12
Genomic Map