Assay Details
| Gene Symbol: Erich6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024300.1 | NP_001019471.1 | 12 - 13 | 1464 | 93 | |
| GenBank mRNA | BC083834.1 | - | 12 - 13 | 1464 | 7112954 | 93 |
More Information
Set Membership: |
|

Erich6
Genomic Map