Assay Details
| Gene Symbol: LOC103691556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761019.1 | XP_008759241.1 | 1361 | 112 | ||
| XM_008761020.1 | XP_008759242.1 | 1361 | 112 | |||
| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138872.1 | NP_620227.1 | 566 | 112 | ||
| XM_008761009.1 | XP_008759231.1 | 1 - 2 | 1361 | 112 | ||
| GenBank mRNA | AF012347.1 | - | 566 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103691556
Genomic Map