Assay Details
| Gene Symbol: Naa15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107674.1 | NP_001101144.1 | 19 - 20 | 2626 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P466316 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P081556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2316 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P466316 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P081556 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P081556 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P466316 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P466316 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P081556 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17266 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P445536 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1071405 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P992502 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72304 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10815301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815297 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815299 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815292 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10815284 | - | - |
| Rat Genome 230 2.0 Array | 1393078_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383514_s_at | - | - |
| Rat Genome 230 2.0 Array | 1372558_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372558_at | - | - |
| Rat Genome 230 2.0 Array | 1377612_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377612_at | - | - |
| Rat Genome 230 2.0 Array | 1393881_at | - | - |
| Rat Genome 230 2.0 Array | 1383514_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1393881_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393078_at | - | - |
| Affymetrix HT Rat Focus Array | 1393078_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372558_at | - | - |
| Affymetrix HT Rat Focus Array | 1372558_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383514_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1393881_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393078_at | - | - |
| Affymetrix HT Rat Focus Array | 1383514_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1377612_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393881_at | - | - |
| Affymetrix HT Rat Focus Array | 1377612_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200007050.rn.1 | TC0200001110.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200007051.rn.1 | TC0200001110.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200007048.rn.1 | TC0200001110.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200007049.rn.1 | TC0200001110.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372558_PM_at | - | - |
| Rat Expression Set 230 | 1372558_at | - | - |
| Rat Expression Set 230 | 1383514_s_at | - | - |
| Rat Expression Set 230 | 1383514_PM_s_at | - | - |
| Rat Expression Set 230 | 1377612_PM_at | - | - |
| Rat Expression Set 230 | 1393078_PM_at | - | - |
| Rat Expression Set 230 | 1393881_at | - | - |
| Rat Expression Set 230 | 1377612_at | - | - |
| Rat Expression Set 230 | 1393078_at | - | - |
| Rat Expression Set 230 | 1393881_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815284 | - | - |
| Rat Genome U34 Set | rc_AI177404_at | - | - |
| Rat Genome U34 Set | rc_AA849957_at | - | - |
| Rat Genome U34 Set | rc_AI179519_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21624528 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352365 | - | - |

Naa15
Genomic Map