Assay Details
| Gene Symbol: Cog6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004262.1 | NP_001004262.1 | 18 - 19 | 1833 | 95 | |
| GenBank mRNA | BC079442.1 | - | 18 - 19 | 1833 | 7115484 | 95 |
| FQ225755.1 | - | 1624 | 95 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P356014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P356014 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P356014 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P356014 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P356014 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16934 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23370 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84195 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10823155 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823165 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823156 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823164 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823171 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823168 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823170 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823157 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823154 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823167 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823153 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823169 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10823152 | - | - |
| Rat Genome 230 2.0 Array | 1398471_at | - | - |
| Rat Genome 230 2.0 Array | 1398471_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200024681.rn.1 | TC0200003672.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398471_PM_at | - | - |
| Rat Expression Set 230 | 1398471_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823152 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21435751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351803 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65560 | - | - |

Cog6
Genomic Map