Assay Details
| Gene Symbol: Gpr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232401.2 | XP_006232463.1 | 1 - 2 | 663 | 159 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232401.2 transcripts.
Set Membership: |
|

Gpr87
Genomic Map