Assay Details
| Gene Symbol: Mbnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191566.1 | NP_001178495.1 | 3 - 4 | 551 | 60 | |
| XM_006232412.2 | XP_006232474.1 | 5 - 6 | 1684 | 60 | ||
| XM_006232430.2 | XP_006232492.1 | 4 - 5 | 1026 | 60 | ||
| XM_006232434.2 | XP_006232496.1 | 3 - 4 | 547 | 60 | ||
| XM_008761032.1 | XP_008759254.1 | 4 - 5 | 678 | 60 | ||
| XM_008761033.1 | XP_008759255.1 | 4 - 5 | 678 | 60 | ||
| XM_008761034.1 | XP_008759256.1 | 4 - 5 | 678 | 60 | ||
| XM_008761035.1 | XP_008759257.1 | 3 - 4 | 567 | 60 | ||
| XM_008761036.1 | XP_008759258.1 | 4 - 5 | 678 | 60 | ||
| XM_008761037.1 | XP_008759259.1 | 4 - 5 | 678 | 60 | ||
| XM_008761038.1 | XP_008759260.1 | 3 - 4 | 567 | 60 | ||
| XM_008761039.1 | XP_008759261.1 | 4 - 5 | 678 | 60 | ||
| XM_008761040.1 | XP_008759262.1 | 3 - 4 | 567 | 60 | ||
| XM_008761042.1 | XP_008759264.1 | 3 - 4 | 567 | 60 | ||
| XM_008761043.1 | XP_008759265.1 | 3 - 4 | 567 | 60 | ||
| XM_008761044.1 | XP_008759266.1 | 3 - 4 | 567 | 60 | ||
| XM_008761045.1 | XP_008759267.1 | 3 - 4 | 567 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbnl1
Genomic Map