Assay Details
| Gene Symbol: Cox7a2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239680.1 | XP_006239742.1 | 1 - 2 | 147 | 78 | |
| GenBank mRNA | BC166896.1 | - | 1 - 2 | 130 | 6921504 | 78 |
| FQ212738.1 | - | 1 - 2 | 96 | 78 | ||
| FQ213890.1 | - | 1 - 2 | 130 | 78 | ||
| FQ215773.1 | - | 1 - 2 | 99 | 78 | ||
| FQ220707.1 | - | 1 - 2 | 130 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox7a2l
Genomic Map