Assay Details
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035221.1 | NP_001030298.1 | 85 | 80 | ||
GenBank mRNA | CB584328.1 | - | 96 | 80 | ||
FQ229713.1 | - | 52 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001035221.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
