Assay Details
| Gene Symbol: Srsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039035.2 | NP_001034124.2 | 2 - 3 | 304 | 99 | |
| XM_006239642.1 | XP_006239704.1 | 2 - 3 | 344 | 99 | ||
| XM_006239643.1 | XP_006239705.1 | 2 - 3 | 344 | 99 | ||
| XM_006239644.1 | XP_006239706.1 | 2 - 3 | 344 | 99 | ||
| XM_006239645.1 | XP_006239707.1 | 2 - 3 | 344 | 99 | ||
| XM_006239646.1 | XP_006239708.1 | 2 - 3 | 344 | 99 | ||
| GenBank mRNA | BC099175.1 | - | 2 - 3 | 294 | 7382701 | 99 |
| FM084661.1 | - | 2 - 3 | 304 | 99 | ||
| FM085512.1 | - | 2 - 3 | 298 | 99 | ||
| FQ229134.1 | - | 2 - 3 | 311 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srsf7
Genomic Map