Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191847.1 | NP_001178776.1 | 2 - 3 | 238 | 64 | |
| XM_008761056.1 | XP_008759278.1 | 2 - 3 | 240 | 64 | ||
| GenBank mRNA | FQ217493.1 | - | 223 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd1
Genomic Map