Assay Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109051.1 | NP_001102521.1 | 1 - 2 | 94 | 149 | |
| XM_006247907.2 | XP_006247969.1 | 3 - 4 | 990 | 149 | ||
| GenBank mRNA | FQ212712.1 | - | 5 - 6 | 555 | 149 | |
| FQ212922.1 | - | 3 - 4 | 458 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fn3k
Genomic Map