Assay Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109051.1 | NP_001102521.1 | 2 - 3 | 178 | 102 | |
| XM_006247907.2 | XP_006247969.1 | 4 - 5 | 1074 | 102 | ||
| GenBank mRNA | FQ212712.1 | - | 6 - 7 | 639 | 102 | |
| FQ212922.1 | - | 4 - 5 | 542 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fn3k
Genomic Map