Assay Details
| Gene Symbol: Fxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100647.1 | NP_001094117.1 | 6 - 7 | 557 | 89 | |
| GenBank mRNA | BC160929.1 | - | 6 - 7 | 557 | 8364923 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Fxr2
Genomic Map