Assay Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019325.1 | NP_062198.1 | 1 - 2 | 58 | 72 | |
| GenBank mRNA | BC113948.1 | - | 1 - 2 | 58 | 7937870 | 72 |
| DQ872907.1 | - | 2 - 3 | 73 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC113948.1, DQ872907.1, NM_019325.1 transcripts.
Set Membership: |
|

Myh4
Genomic Map