Assay Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019325.1 | NP_062198.1 | 33 - 34 | 5062 | 65 | |
| GenBank mRNA | BC113948.1 | - | 33 - 34 | 5062 | 7937870 | 65 |
| FQ215406.1 | - | 2 - 3 | 370 | 65 | ||
| FQ216538.1 | - | 2 - 3 | 354 | 65 | ||
| FQ224129.1 | - | 9 - 10 | 1917 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50636 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071692 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071692 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071692 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P708941 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63212 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10734587 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734608 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734611 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734592 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734605 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734586 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734600 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734589 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734584 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734597 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734599 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734594 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734591 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734598 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734601 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734606 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734588 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734585 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734596 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734590 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734595 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734603 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734593 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734614 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10734583 | - | - |
| Rat Genome 230 2.0 Array | 1370900_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370900_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000007353.rn.1 | TC1000000864.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000007351.rn.1 | TC1000000864.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000007350.rn.1 | TC1000000864.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000007354.rn.1 | TC1000000864.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000007355.rn.1 | TC1000000864.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000007352.rn.1 | TC1000000864.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370900_PM_at | - | - |
| Rat Expression Set 230 | 1370900_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10734583 | - | - |
| Rat Genome U34 Set | L24897_s_at | - | - |
| Rat Genome U34 Set | rc_AI010562_at | - | - |
| Rat Genome U34 Set | rc_AI172178_i_at | - | - |
| Rat Genome U34 Set | rc_AI175304_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21544903 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374894 | - | - |

Myh4
Genomic Map