Assay Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019325.1 | NP_062198.1 | 2 - 3 | 298 | 120 | |
| GenBank mRNA | BC113948.1 | - | 2 - 3 | 298 | 7937870 | 120 |
| DQ872907.1 | - | 3 - 4 | 313 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myh4
Genomic Map