Assay Details
| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105774.1 | NP_001099244.1 | 2 - 3 | 315 | 102 | |
| XM_006245937.2 | XP_006245999.1 | 2 - 3 | 370 | 102 | ||
| GenBank mRNA | BC161966.1 | - | 2 - 3 | 313 | 9028415 | 102 |
| FQ213277.1 | - | 2 - 3 | 236 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpusd1
Genomic Map