Assay Details
| Gene Symbol: Ace3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258237.1 | NP_001245166.1 | 13 - 14 | 1858 | 158 | |
| GenBank mRNA | BC085122.1 | - | 12 - 13 | 1950 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ace3
Genomic Map