Assay Details
| Gene Symbol: Mpdu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107011.1 | NP_001100481.1 | 1 - 2 | 138 | 104 | |
| GenBank mRNA | BC087071.1 | - | 1 - 2 | 284 | 104 | |
| BC107940.1 | - | 1 - 2 | 97 | 104 | ||
| FQ213141.1 | - | 1 - 2 | 114 | 104 | ||
| FQ220130.1 | - | 1 - 2 | 102 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpdu1
Genomic Map