Assay Details
| Gene Symbol: Mpdu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107011.1 | NP_001100481.1 | 2 - 3 | 197 | 115 | |
| GenBank mRNA | BC107940.1 | - | 2 - 3 | 156 | 115 | |
| FQ213141.1 | - | 2 - 3 | 173 | 115 | ||
| FQ220130.1 | - | 2 - 3 | 161 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpdu1
Genomic Map