Assay Details
| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021594.1 | NP_067605.1 | 2 - 3 | 663 | 84 | |
| GenBank mRNA | AF154336.1 | - | 2 - 3 | 663 | 84 | |
| FQ213792.1 | - | 2 - 3 | 780 | 84 | ||
| FQ228926.1 | - | 2 - 3 | 761 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r1
Genomic Map