Assay Details
| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021594.1 | NP_067605.1 | 3 - 4 | 815 | 133 | |
| GenBank mRNA | AF154336.1 | - | 3 - 4 | 815 | 133 | |
| FQ213792.1 | - | 3 - 4 | 932 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r1
Genomic Map