Assay Details
| Gene Symbol: Sh3bp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127581.1 | NP_001121053.1 | 3 - 4 | 819 | 88 | |
| XM_008767708.1 | XP_008765930.1 | 4 - 5 | 936 | 88 | ||
| XM_008767709.1 | XP_008765931.1 | 4 - 5 | 906 | 88 | ||
| XM_008767710.1 | XP_008765932.1 | 4 - 5 | 944 | 88 | ||
| GenBank mRNA | BC166447.1 | - | 3 - 4 | 819 | 8364461 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bp5l
Genomic Map