Assay Details
| Gene Symbol: Efcab5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246915.2 | XP_006246977.1 | 15 - 16 | 3266 | 62 | |
| XM_006246917.2 | XP_006246979.1 | 13 - 14 | 3049 | 62 | ||
| XM_008768139.1 | XP_008766361.1 | 11 - 12 | 2473 | 62 | ||
More Information
Set Membership: |
|

Efcab5
Genomic Map