Assay Details
| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008761.1 | NP_001008761.1 | 1 - 2 | 533 | 66 | |
| XM_006247357.2 | XP_006247419.1 | 1 - 2 | 1464 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt12
Genomic Map