Assay Details
| Gene Symbol: Tp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105818.1 | NP_001099288.1 | 3 - 4 | 247 | 125 | |
| GenBank mRNA | BC158678.1 | - | 3 - 4 | 209 | 8372780 | 125 |
More Information
Updated Mapping Information
Set Membership: |
|

Tp53i13
Genomic Map