Assay Details
| Gene Symbol: Tp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105818.1 | NP_001099288.1 | 5 - 6 | 589 | 112 | |
| XM_006246902.2 | XP_006246964.1 | 4 - 5 | 616 | 112 | ||
| GenBank mRNA | BC158678.1 | - | 5 - 6 | 551 | 8372780 | 112 |
More Information
Updated Mapping Information
Set Membership: |
|

Tp53i13
Genomic Map