Assay Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108818.1 | NP_001102288.1 | 1 - 2 | 684 | 59 | |
| XM_008773349.1 | XP_008771571.1 | 1 - 2 | 107 | 59 | ||
| GenBank mRNA | BC166918.1 | - | 1 - 2 | 47 | 59 | |
| FQ220252.1 | - | 1 - 2 | 48 | 59 | ||
| FQ225636.1 | - | 1 - 2 | 67 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pbdc1
Genomic Map