Assay Details
| Gene Symbol: LOC100910646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752081.3 | XP_003752129.1 | 5620 | 111 | ||
| XM_006257141.2 | XP_006257203.1 | 5732 | 111 | |||
| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257149.2 | XP_006257211.1 | 6 | 5732 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910646
Genomic Map