Assay Details
| Gene Symbol: Rab7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109328.2 | NP_001102798.1 | 1 - 2 | 96 | 90 | |
| XM_008769490.1 | XP_008767712.1 | 1 - 2 | 165 | 90 | ||
| GenBank mRNA | FM076322.1 | - | 1 - 2 | 96 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109328.2, XM_008769490.1 transcripts.
Set Membership: |
|

Rab7b
Genomic Map