Assay Details
| Gene Symbol: Rab7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109328.2 | NP_001102798.1 | 2 - 3 | 159 | 116 | |
| XM_008769490.1 | XP_008767712.1 | 2 - 3 | 228 | 116 | ||
| XM_008769491.1 | XP_008767713.1 | 2 - 3 | 574 | 116 | ||
| XM_008769492.1 | XP_008767714.1 | 2 - 3 | 575 | 116 | ||
| XM_008769493.1 | XP_008767715.1 | 2 - 3 | 579 | 116 | ||
| GenBank mRNA | FM076322.1 | - | 2 - 3 | 159 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab7b
Genomic Map