Assay Details
Gene Symbol: RGD1564614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134792.1 | NP_001128264.1 | 5 - 6 | 824 | 140 | |
XM_006249945.2 | XP_006250007.1 | 6 - 7 | 1047 | 140 | ||
XM_006249946.2 | XP_006250008.1 | 4 - 5 | 675 | 140 | ||
XM_008769568.1 | XP_008767790.1 | 6 - 7 | 1047 | 140 | ||
XM_008769569.1 | XP_008767791.1 | 6 - 7 | 963 | 140 | ||
GenBank mRNA | AF436847.2 | - | 5 - 6 | 824 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
