Assay Details
| Gene Symbol: Angptl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109383.1 | NP_001102853.1 | 1 - 2 | 289 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109383.1 transcripts.
Set Membership: |
|

Angptl1
Genomic Map