Assay Details
| Gene Symbol: Cpeb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760379.1 | XP_008758601.1 | 5 - 6 | 2753 | 92 | |
| XM_008760380.1 | XP_008758602.1 | 5 - 6 | 2684 | 92 | ||
| XM_008760381.1 | XP_008758603.1 | 5 - 6 | 2245 | 92 | ||
| XM_008760382.1 | XP_008758604.1 | 4 - 5 | 1464 | 92 | ||
| XM_008760383.1 | XP_008758605.1 | 5 - 6 | 1599 | 92 | ||
| XM_008760384.1 | XP_008758606.1 | 5 - 6 | 2753 | 92 | ||
More Information
Set Membership: |
|

Cpeb3
Genomic Map