Assay Details
| Gene Symbol: Rab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109979.2 | NP_001103449.1 | 260 | 73 | ||
| GenBank mRNA | BC085118.1 | - | 227 | 73 | ||
| DN934711.1 | - | 278 | 73 | |||
| EV762710.1 | - | 279 | 73 | |||
| Gene Symbol: LOC102555167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760197.1 | XP_008758419.1 | 1 | 228 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rab1b
Genomic Map