Assay Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107513.1 | NP_001100983.1 | 1 - 2 | 130 | 139 | |
| XM_006229089.2 | XP_006229151.1 | 1 - 2 | 125 | 139 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107513.1,XM_006229089.2 transcripts.
Set Membership: |
|

Tmem143
Genomic Map