Assay Details
| Gene Symbol: Smim11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131000.1 | NP_001124472.1 | 2 - 3 | 157 | 86 | |
| XM_006248022.1 | XP_006248084.1 | 2 - 3 | 221 | 86 | ||
| GenBank mRNA | BC166910.1 | - | 2 - 3 | 158 | 6890105 | 86 |
| BC166926.1 | - | 2 - 3 | 158 | 6919678 | 86 | |
| FQ212222.1 | - | 2 - 3 | 187 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166910.1, BC166926.1, NM_001131000.1, XM_006248022.1 transcripts.
Set Membership: |
|

Smim11
Genomic Map