Assay Details
| Gene Symbol: Tomm20l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001072851.4 | XP_001072851.3 | 1 - 2 | 583 | 104 | |
| XM_008764723.1 | XP_008762945.1 | 1 - 2 | 583 | 104 | ||
More Information
Set Membership: |
|

Tomm20l
Genomic Map