Assay Details
| Gene Symbol: Tomm20l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001072851.4 | XP_001072851.3 | 2 - 3 | 623 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm20l| Gene Symbol: Tomm20l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001072851.4 | XP_001072851.3 | 2 - 3 | 623 | 120 | |
Set Membership: |
|