Assay Details
| Gene Symbol: Sgpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_343081.7 | XP_343082.4 | 2 - 3 | 877 | 130 | |
| GenBank mRNA | AF329638.1 | - | 2 - 3 | 375 | 130 | |
More Information
Set Membership: |
|

Sgpp1
Genomic Map