Assay Details
| Gene Symbol: Rdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012193.1 | NP_001012193.1 | 3 - 4 | 419 | 92 | |
| GenBank mRNA | AY387097.1 | - | 3 - 4 | 587 | 92 | |
| BC079276.1 | - | 3 - 4 | 419 | 7134077 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rdh11
Genomic Map