Assay Details
| Gene Symbol: Asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009716.1 | NP_001009716.1 | 1 - 2 | 261 | 67 | |
| GenBank mRNA | BC088863.1 | - | 1 - 2 | 261 | 7313227 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088863.1, NM_001009716.1 transcripts.
Set Membership: |
|

Asphd2
Genomic Map