Assay Details
| Gene Symbol: Ran | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249285.2 | XP_006249347.1 | 1 - 2 | 54 | 217 | |
| GenBank mRNA | BC059123.1 | - | 1 - 2 | 82 | 6920235 | 217 |
| FQ220083.1 | - | 1 - 2 | 50 | 217 | ||
| FQ220326.1 | - | 1 - 2 | 50 | 217 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059123.1, XM_006249285.2 transcripts.
Set Membership: |
|

Ran
Genomic Map