Assay Details
| Gene Symbol: Oas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144752.2 | NP_653353.2 | 1 - 2 | 205 | 86 | |
| GenBank mRNA | AF068268.1 | - | 217 | 86 | ||
| BC091121.1 | - | 1 - 2 | 205 | 7383389 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oas1b
Genomic Map