Assay Details
| Gene Symbol: Oas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144752.2 | NP_653353.2 | 2 - 3 | 493 | 79 | |
| GenBank mRNA | BC091121.1 | - | 2 - 3 | 493 | 7383389 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|

Oas1b
Genomic Map