Assay Details
Gene Symbol: Fam109a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727967.3 | XP_002728013.1 | 3 - 4 | 323 | 91 | |
XM_006249410.2 | XP_006249472.1 | 2 - 3 | 240 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002727967.3, XM_006249410.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
