Assay Details
| Gene Symbol: Pebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017236.1 | NP_058932.1 | 2 - 3 | 300 | 117 | |
| GenBank mRNA | BC063171.1 | - | 2 - 3 | 317 | 6888051 | 117 |
| FQ212424.1 | - | 2 - 3 | 283 | 117 | ||
| FQ215504.1 | - | 2 - 3 | 311 | 117 | ||
| FQ224857.1 | - | 2 - 3 | 311 | 117 | ||
| X71873.1 | - | 2 - 3 | 275 | 117 | ||
| X75253.1 | - | 2 - 3 | 300 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pebp1
Genomic Map